2,6-Dimethylbenzeneboronic acid, 97%
2,6-Dimethylbenzeneboronic acid, 97%
2,6-Dimethylbenzeneboronic acid, 97%
Thermo Scientific Chemicals

2,6-Dimethylbenzeneboronic acid, 97%

CAS: 100379-00-8 | C8H11BO2 | 149.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24613.14
also known as B24613-14
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
297.65
Online Exclusive
331.00
Save 33.35 (10%)
Each
Chemical Identifiers
CAS100379-00-8
IUPAC Name(2,6-dimethylphenyl)boronic acid
Molecular FormulaC8H11BO2
InChI KeyZXDTWWZIHJEZOG-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMRConforms to structure
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Linglin Wu; Emma Drinkel; Fiona Gaggia; Samanta Capolicchio; Anthony Linden; Laura Falivene; Luigi Cavallo; Reto Dorta. Room-temperature synthesis of tetra-ortho-substituted biaryls by NHC-catalyzed Suzuki-Miyaura couplings. Chemistry: A European Journal. 2011, 17 (46), 12886-12890.
  2. Srimanta Manna; Soham Maity; Sujoy Rana; Soumitra Agasti; Debabrata Maiti. ipso-Nitration of arylboronic acids with bismuth nitrate and perdisulfate. Organic Letters. 2012, 14 (7), 1736-1739.