2,4-Dimethoxybenzonitrile, 99%
2,4-Dimethoxybenzonitrile, 99%
2,4-Dimethoxybenzonitrile, 99%
2,4-Dimethoxybenzonitrile, 99%
Thermo Scientific Chemicals

2,4-Dimethoxybenzonitrile, 99%

CAS: 4107-65-7 | C9H9NO2 | 163.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24621.14
also known as B24621-14
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.50
Save 7.85 (16%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.50
Save 7.85 (16%)
Each
Add to cart
Chemical Identifiers
CAS4107-65-7
IUPAC Name2,4-dimethoxybenzonitrile
Molecular FormulaC9H9NO2
InChI KeyRYRZSQQELLQCMZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C#N)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Melting Point (clear melt)89.5-95.5?C
Assay (GC)≥98.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only