tert-Pentylamine, 98%
tert-Pentylamine, 98%
tert-Pentylamine, 98%
Thermo Scientific Chemicals

tert-Pentylamine, 98%

CAS: 594-39-8 | C5H13N | 87.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24639.14
also known as B24639-14
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.30
Save 11.65 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 30-Jun-2026
78.30
Save 11.65 (15%)
Each
Chemical Identifiers
CAS594-39-8
IUPAC Name2-methylbutan-2-aminium
Molecular FormulaC5H14N
InChI KeyGELMWIVBBPAMIO-UHFFFAOYSA-O
SMILESCCC(C)(C)[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.3975-1.4015 @ 20°C
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only