tert-Pentylamine, 98%
tert-Pentylamine, 98%
tert-Pentylamine, 98%
tert-Pentylamine, 98%
Thermo Scientific Chemicals

tert-Pentylamine, 98%

CAS: 594-39-8 | C5H13N | 87.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B24639.22
also known as B24639-22
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS594-39-8
IUPAC Name2-methylbutan-2-aminium
Molecular FormulaC5H14N
InChI KeyGELMWIVBBPAMIO-UHFFFAOYSA-O
SMILESCCC(C)(C)[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.3975-1.4015 @ 20°C
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only