1-(2-Aminoethyl)pyrrolidine, 99%
1-(2-Aminoethyl)pyrrolidine, 99%
1-(2-Aminoethyl)pyrrolidine, 99%
Thermo Scientific Chemicals

1-(2-Aminoethyl)pyrrolidine, 99%

CAS: 7154-73-6 | C6H14N2 | 114.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24645.06
also known as B24645-06
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Chemical Identifiers
CAS7154-73-6
IUPAC Name2-(pyrrolidin-1-yl)ethan-1-amine
Molecular FormulaC6H14N2
InChI KeyWRXNJTBODVGDRY-UHFFFAOYSA-N
SMILESNCCN1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.4675-1.4705 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only