3,5-Dichlorobenzoyl chloride, 96%
3,5-Dichlorobenzoyl chloride, 96%
3,5-Dichlorobenzoyl chloride, 96%
3,5-Dichlorobenzoyl chloride, 96%
Thermo Scientific Chemicals

3,5-Dichlorobenzoyl chloride, 96%

CAS: 2905-62-6 | C7H3Cl3O | 209.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24676.14
also known as B24676-14
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
133.65
Online Exclusive
148.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS2905-62-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5790-1.5850 @ 20°C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only