(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%
Thermo Scientific Chemicals

(R)-(+)-2-Pyrrolidinone-5-carboxylic acid, 98+%

CAS: 4042-36-8 | C5H7NO3 | 129.115 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24678.22
also known as B24678-22
Price (USD)
858.65
Online Exclusive
954.00
Save 95.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
858.65
Online Exclusive
954.00
Save 95.35 (10%)
Each
Chemical Identifiers
CAS4042-36-8
IUPAC Name5-oxopyrrolidine-2-carboxylic acid
Molecular FormulaC5H7NO3
InChI KeyODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILESOC(=O)C1CCC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)151-163?C
Optical Rotation+10.5? ? 1? (c=5 in water)
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Pyrrolidinone-5-carboxylic acid serves as a building block in the synthesis of diphthamide.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing /reducing agents, strong acids, strong bases.
RUO – Research Use Only

General References:

  1. Bing Yan; Hai-Feng Lu. Lanthanide-centered inorganic/organic hybrids from functionalized 2-pyrrolidinone-5-carboxylic acid bridge: Covalently bonded assembly and luminescence. Journal of Organometallic Chemistry. 2009, 694 (16), 2597-2603.
  2. Masahito Ochiai, Minako Inenaga; Yoshimitsu Nagao Robert M. Moriarty; Radhe K Vaid; Michael P. Duncan. Oxidative decarboxylation of cyclic amino acids and dehydrogenation of cyclic secondary amines with iodosobenzene. Tetrahedron Letters. 1988, 29 (52), 6917-6920.
  3. For applications, see next entry.