Aminoguanidine hydrogen carbonate, 98+%
Aminoguanidine hydrogen carbonate, 98+%
Aminoguanidine hydrogen carbonate, 98+%
Thermo Scientific Chemicals

Aminoguanidine hydrogen carbonate, 98+%

CAS: 2582-30-1 | C2H8N4O3 | 136.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number B24696.0E
also known as B24696-0E
Price (USD)
163.65
Special offer
Online exclusive
Ends: 30-Jun-2026
192.00
Save 28.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
163.65
Special offer
Online exclusive
Ends: 30-Jun-2026
192.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS2582-30-1
IUPAC NameN''-aminoguanidine; carbonic acid
Molecular FormulaC2H8N4O3
InChI KeyOTXHZHQQWQTQMW-UHFFFAOYSA-N
SMILESOC(O)=O.NN=C(N)N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine bicarbonate is an inhibitor of NOS (nitric oxide synthase). Aminoguanidine bicarbonate is used to study the effect of addition of polyamines to rat embryo cell cultures infected with adenovirus type 5.

Solubility
Soluble in water (3.3 mg/ml at 30°C), and water (2.7 mg/ml at 20°C).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. T. George.; C. L. Kaul.; R. S. Grewal.; R. Tahilramani. Antihypertensive and monoamine oxidize inhibitory activity of some derivatives of 3-formyl-4-oxo-4H-pyrido[1,2-a]pyrimidine. J. Med. Chem. 1971, 14, (10), 913-915.
  2. Jaroslav Stanek.; Giorgio Caravatti.; Jorg Frei.; Pascal Furet.; Helmut Mett.; Peter Schneider.; Urs Regenass. 4-Amidinoindan-1-one 2'-amidinohydrazone: a new potent and selective inhibitor of S-adenosylmethionine decarboxylase. J. Med. Chem. 1993, 36, (15), 2168-2171.