3,4-Dichlorobenzaldehyde, 97%
3,4-Dichlorobenzaldehyde, 97%
3,4-Dichlorobenzaldehyde, 97%
Thermo Scientific Chemicals

3,4-Dichlorobenzaldehyde, 97%

CAS: 6287-38-3 | C7H4Cl2O | 175.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24706.14
also known as B24706-14
Price (USD)
63.65
Online Exclusive
70.10
Save 6.45 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
63.65
Online Exclusive
70.10
Save 6.45 (9%)
Each
Chemical Identifiers
CAS6287-38-3
IUPAC Name3,4-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyZWUSBSHBFFPRNE-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Identification (FTIR)Conforms
Free acid (titration)≤1.5%
Melting Point (clear melt)40-45?C
Appearance (Color)White to pale yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only