2,2'-Thenil, 98%
2,2'-Thenil, 98%
2,2'-Thenil, 98%
Thermo Scientific Chemicals

2,2'-Thenil, 98%

CAS: 7333-07-5 | C10H6O2S2 | 222.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24753.06
also known as B24753-06
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS7333-07-5
IUPAC Namebis(thiophen-2-yl)ethane-1,2-dione
Molecular FormulaC10H6O2S2
InChI KeyUNWKVSDABPCZMK-UHFFFAOYSA-N
SMILESO=C(C(=O)C1=CC=CS1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)81-85?C
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Identification (FTIR)Conforms
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thenil is a non-steroidal anti-inflammatory agent and also used for pain and inflammation in muscles and joints.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. E. Thomas Strom.; G. A. Russell.; Joseph H. Schoeb. Semidiones. II. Radical Anions of 2,2'-Furil, 2,2'-Thenil, and 3,3'-Thenil1. J. Am. Chem. Soc. 1966, 88, (9), 2004-2007.
  2. Jean Sice. Preparation and Reactions of 2-Methoxythiophene. J. Am. Chem. Soc. 1953, 75, (15), 3697-3700.