3-Ethoxy-4-methoxybenzaldehyde, 99%
3-Ethoxy-4-methoxybenzaldehyde, 99%
3-Ethoxy-4-methoxybenzaldehyde, 99%
3-Ethoxy-4-methoxybenzaldehyde, 99%
Thermo Scientific Chemicals

3-Ethoxy-4-methoxybenzaldehyde, 99%

CAS: 1131-52-8 | C10H12O3 | 180.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24762.14
also known as B24762-14
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1131-52-8
IUPAC Name3-ethoxy-4-methoxybenzaldehyde
Molecular FormulaC10H12O3
InChI KeyVAMZHXWLGRQSJS-UHFFFAOYSA-N
SMILESCCOC1=CC(C=O)=CC=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥98.5%
Free acid (titration)≤1.0%
Melting Point (clear melt)47.0-54.0°C
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-4-methoxybenzaldehyde is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Store in a tightly sealed container. Store away from oxidizing agents, air.
RUO – Research Use Only

General References:

  1. E. Kiehlmann; J. E. Conn & J. H. Borden. 7-Ethoxy-6-Methoxy-2,2-Dimethyl-2H-1-Benzopyran. Organic Preparations and Procedures International: The New Journal for Organic Synthesis. 1982, 14 (5), 337-342.
  2. Alfred Burger; Richard D. Foggio. Nuclear Substituted 3,4-Dihydroxyphenethylamines and Related Derivatives. J. Am. Chem. Soc. 1956, 78 (17), 4419-4422.