Ethyl hippurate, 96%
Ethyl hippurate, 96%
Ethyl hippurate, 96%
Thermo Scientific Chemicals

Ethyl hippurate, 96%

CAS: 1499-53-2 | C11H13NO3 | 207.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24765.14
also known as B24765-14
Price (USD)
192.65
Special offer
Online exclusive
Ends: 30-Jun-2026
226.00
Save 33.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
192.65
Special offer
Online exclusive
Ends: 30-Jun-2026
226.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS1499-53-2
IUPAC Nameethyl 2-(phenylformamido)acetate
Molecular FormulaC11H13NO3
InChI KeyPTXRQIPIELXJFH-UHFFFAOYSA-N
SMILESCCOC(=O)CNC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)56.0-62.0°C
Appearance (Color)White to pale cream
Assay (GC)≥95.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only