2,5-Dichlorophenol, 98%
2,5-Dichlorophenol, 98%
2,5-Dichlorophenol, 98%
Thermo Scientific Chemicals

2,5-Dichlorophenol, 98%

CAS: 583-78-8 | C6H4Cl2O | 163.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B24787.36
also known as B24787-36
Price (USD)
312.65
Special offer
Online exclusive
Ends: 30-Jun-2026
368.00
Save 55.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
312.65
Special offer
Online exclusive
Ends: 30-Jun-2026
368.00
Save 55.35 (15%)
Each
Chemical Identifiers
CAS583-78-8
IUPAC Name2,5-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyRANCECPPZPIPNO-UHFFFAOYSA-N
SMILESOC1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)53.0-60.0?C
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only