2,4,6-Trimethylthiophenol, 97%
2,4,6-Trimethylthiophenol, 97%
2,4,6-Trimethylthiophenol, 97%
2,4,6-Trimethylthiophenol, 97%
Thermo Scientific Chemicals

2,4,6-Trimethylthiophenol, 97%

CAS: 1541-10-2 | C9H12S | 152.255 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24798.06
also known as B24798-06
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1541-10-2
IUPAC Name(2,4,6-trimethylphenyl)sulfanide
Molecular FormulaC9H11S
InChI KeyPKANQZUPJCMBAK-UHFFFAOYSA-M
SMILESCC1=CC(C)=C([S-])C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥96.0%
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Kenzo Tanaka.; Atsunori Mori.; Shohei Inoue. The cyclic dipeptide cyclo[(S)-phenylalanyl-(S)-histidyl] as a catalyst for asymmetric addition of hydrogen cyanide to aldehydes. J. Org. Chem. 1990, 55, (1), 181-185.
  2. Ichiro Mori.; Paul A. Bartlett.; Clayton H. Heathcock. High diastereofacial selectivity in nucleophilic additions to chiral thionium ions. J. Am. Chem. Soc. 1987, 109, (23), 7199-7200.