1-(2-Chlorophenyl)ethanol, 96%
1-(2-Chlorophenyl)ethanol, 96%
1-(2-Chlorophenyl)ethanol, 96%
Thermo Scientific Chemicals

1-(2-Chlorophenyl)ethanol, 96%

CAS: 13524-04-4 | C8H9ClO | 156.609 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24800.14
also known as B24800-14
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS13524-04-4
IUPAC Name1-(2-chlorophenyl)ethan-1-ol
Molecular FormulaC8H9ClO
InChI KeyDDUBOVLGCYUYFX-UHFFFAOYNA-N
SMILESCC(O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5420-1.5480 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to orange
Assay (GC)≥95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only