2,4,6-Triphenylaniline, 98%
2,4,6-Triphenylaniline, 98%
2,4,6-Triphenylaniline, 98%
Thermo Scientific Chemicals

2,4,6-Triphenylaniline, 98%

CAS: 6864-20-6 | C24H19N | 321.423 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24871.14
also known as B24871-14
Price (USD)
990.65
Online Exclusive
1,100.00
Save 109.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
990.65
Online Exclusive
1,100.00
Save 109.35 (10%)
Each
Chemical Identifiers
CAS6864-20-6
IUPAC Name3,5-diphenyl-[1,1'-biphenyl]-2-amine
Molecular FormulaC24H19N
InChI KeyAGWKGKUGJDMKMT-UHFFFAOYSA-N
SMILESNC1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)134.0-140.0?C
Appearance (Color)Pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only