5-Bromo-1,2,3-trimethoxybenzene, 97%
5-Bromo-1,2,3-trimethoxybenzene, 97%
5-Bromo-1,2,3-trimethoxybenzene, 97%
Thermo Scientific Chemicals

5-Bromo-1,2,3-trimethoxybenzene, 97%

CAS: 2675-79-8 | C9H11BrO3 | 247.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B24872.06
also known as B24872-06
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.70
Save 7.05 (16%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
Ends: 30-Jun-2026
44.70
Save 7.05 (16%)
Each
Chemical Identifiers
CAS2675-79-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to orange to brown
FormPowder or crystalline powder
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)75.5-84.5°C
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-1,2,3-trimethoxybenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only