1-Thionaphthol, 99%
1-Thionaphthol, 99%
1-Thionaphthol, 99%
Thermo Scientific Chemicals

1-Thionaphthol, 99%

CAS: 529-36-2 | C10H7S | 159.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24876.03
also known as B24876-03
Price (USD)
59.65
Online Exclusive
65.60
Save 5.95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
59.65
Online Exclusive
65.60
Save 5.95 (9%)
Each
Chemical Identifiers
CAS529-36-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.5%
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Thionaphthol is used to produce the di-[1]naphthyl disulfide. It is also used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in diethyl ether and ethanol. Slightly soluble in water and dilute alkalis.

Notes
Store away from air. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only
Xuhong Qian; Tian-Bao Huang; Dong-zhi Wei; Dong-Hui Zhu; Zhub and Wei Yao. Interaction of naphthyl heterocycles with DNA: effects of thiono and thio groups. J. Chem. Soc., Perkin Trans. 2.2000, 715-718.