2,6-Bis(p-tolyl)pyridine, 97%
2,6-Bis(p-tolyl)pyridine, 97%
2,6-Bis(p-tolyl)pyridine, 97%
Thermo Scientific Chemicals

2,6-Bis(p-tolyl)pyridine, 97%

CAS: 14435-88-2 | C19H17N | 259.352 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24894.06
also known as B24894-06
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Specifications
CAS14435-88-2
Chemical Name or Material2,6-Bis(p-tolyl)pyridine
EINECS Number238-409-3
Formula Weight259.35
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only