2-Bromo-5-fluoroaniline, 98%
2-Bromo-5-fluoroaniline, 98%
2-Bromo-5-fluoroaniline, 98%
Thermo Scientific Chemicals

2-Bromo-5-fluoroaniline, 98%

CAS: 1003-99-2 | C6H5BrFN | 190.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24901.06
also known as B24901-06
Price (USD)
53.65
Online Exclusive
59.60
Save 5.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
59.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS1003-99-2
IUPAC Name2-bromo-5-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyFWTXFEKVHSFTDQ-UHFFFAOYSA-N
SMILESNC1=CC(F)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)40.0-47.0°C
Appearance (Color)White to cream to dark brown
FormCrystals or powder or crystalline powder or lumps or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only