(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
(S)-(+)-2-Amino-1-propanol, 98%
Thermo Scientific Chemicals

(S)-(+)-2-Amino-1-propanol, 98%

CAS: 2749-11-3 | C3H10NO | 76.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B24916.06
also known as B24916-06
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS2749-11-3
IUPAC Name(2S)-1-hydroxypropan-2-aminium
Molecular FormulaC3H10NO
InChI KeyBKMMTJMQCTUHRP-VKHMYHEASA-O
SMILESC[C@H]([NH3+])CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Water Content (Karl Fischer Titration)≤1.5%
Optical Rotation22.0 ± 2.0? (C=2 in ethanol)
Refractive Index1.4475-1.4515 @ 20?C
Appearance (Color)Clear colorless to pale yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only