3,4-Dimethylbenzyl alcohol, 97%
Thermo Scientific Chemicals

3,4-Dimethylbenzyl alcohol, 97%

CAS: 6966-10-5 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24926.14
also known as B24926-14
Price (USD)
460.65
Online Exclusive
512.00
Save 51.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
460.65
Online Exclusive
512.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS6966-10-5
IUPAC Name(3,4-dimethylphenyl)methanol
Molecular FormulaC9H12O
InChI KeyOKGZCXPDJKKZAP-UHFFFAOYSA-N
SMILESCC1=CC=C(CO)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)54.0-64.0?C
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only