5-(4-Morpholinyl)-2-nitrophenol, 97%
5-(4-Morpholinyl)-2-nitrophenol, 97%
5-(4-Morpholinyl)-2-nitrophenol, 97%
Thermo Scientific Chemicals

5-(4-Morpholinyl)-2-nitrophenol, 97%

CAS: 175135-19-0 | C10H12N2O4 | 224.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B24950.03
also known as B24950-03
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.70
Save 8.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.70
Save 8.05 (15%)
Each
Chemical Identifiers
CAS175135-19-0
IUPAC Name5-(morpholin-4-yl)-2-nitrobenzen-1-olate
Molecular FormulaC10H11N2O4
InChI KeyXXIIDARBNMZFEH-UHFFFAOYSA-M
SMILES[O-]C1=CC(=CC=C1[N+]([O-])=O)N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale orange
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)139.5-148.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only