2-Phenylmalonamide, 97%
2-Phenylmalonamide, 97%
2-Phenylmalonamide, 97%
Thermo Scientific Chemicals

2-Phenylmalonamide, 97%

CAS: 10255-95-5 | C9H10N2O2 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24972.03
also known as B24972-03
Price (USD)
18.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
18.00
Each
Chemical Identifiers
CAS10255-95-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only