Thermo Scientific Chemicals

4-Fluorosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24992.06
5 g, Each
Thermo Scientific Chemicals

4-Fluorosalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B24992.06
5 g, Each
Quantity
Catalog number: B24992.06
also known as B24992-06
Price (USD)
Price: 184.00
Online price: 158.65
Your price:
Quantity
-

Chemical Identifiers

CAS
345-29-9
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
185.0-191.0?C
Appearance (Color)
White to cream to pale brown
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text