4-Fluorosalicylic acid, 98%
4-Fluorosalicylic acid, 98%
4-Fluorosalicylic acid, 98%
Thermo Scientific Chemicals

4-Fluorosalicylic acid, 98%

CAS: 345-29-9 | C7H5FO3 | 156.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24992.14
also known as B24992-14
Price (USD)
659.65
Online Exclusive
733.00
Save 73.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
659.65
Online Exclusive
733.00
Save 73.35 (10%)
Each
Chemical Identifiers
CAS345-29-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)185.0-191.0?C
Appearance (Color)White to cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorosalicylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Terézia Szabó-Plánka; Ján Moncol; Eszter Tóth; Béla Gyurcsik; Nóra Veronika Nagy; Zuzana Vasková; Antal Rockenbauer; Dušan Valigura. ESR and pH-potentiometric study of the mixed-ligand complex formation in the copper(II)-4-fluorosalicylic acid-N,N-diethylnicotinamide system: Structure and spectral properties of [Cu(4-fluorosalicylate)2(N,N-diethylnicotinamide)2(H2O)2] complex. Polyhedron.2011, 30, 2421-2429.