2-Fluorothiophenol, 97%
2-Fluorothiophenol, 97%
2-Fluorothiophenol, 97%
Thermo Scientific Chemicals

2-Fluorothiophenol, 97%

CAS: 2557-78-0 | C6H4FS | 127.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B24995.14
also known as B24995-14
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
231.65
Online Exclusive
257.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS2557-78-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5530-1.5570
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)>96.0%
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only