Creatine monohydrate, 99%
Creatine monohydrate, 99%
Creatine monohydrate, 99%
Thermo Scientific Chemicals

Creatine monohydrate, 99%

CAS: 6020-87-7 | C4H11N3O3 | 149.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number B25009.0E
also known as B25009-0E
Price (USD)
-
Quantity:
2500 g
Request bulk or custom format
Chemical Identifiers
CAS6020-87-7
IUPAC Name2-(N-methylcarbamimidamido)acetic acid hydrate
Molecular FormulaC4H11N3O3
InChI KeyMEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILESO.CN(CC(O)=O)C(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder
Loss on Drying6.43-17.09% (105°C, 0.5-1.5 waters)
Appearance (Color)White to cream to yellow
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Creatine monohydrate is involved in rapid ATP production primarily in skeletal muscle tissue via the action of creatine kinase(s). It may be used as a supplement to study its uptake mechanism and metabolism of action. It is used in the treatment of neuromuscular diseases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (17 g/L) at 20°C. Insoluble in ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. Clare E Turner; Winston D Byblow; Nicholas Gant. Creatine supplementation enhances corticomotor excitability and cognitive performance during oxygen deprivation. Journal of Neuroscience.2015, 35, (4), 1773-1780.
  2. Mark D Schuenke; Naomi E Brooks; Robert S Hikida. Interactions of aging, overload, and creatine supplementation in rat plantaris muscle. Journal of Aging Research.2011, 2011, 393416.