3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
3,4-Dimethoxybenzylamine, 97%
Thermo Scientific Chemicals

3,4-Dimethoxybenzylamine, 97%

CAS: 5763-61-1 | C9H13NO2 | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Catalog number B25033.22
also known as B25033-22
Price (USD)
1,013.65
Online Exclusive
1,126.00
Save 112.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
1,013.65
Online Exclusive
1,126.00
Save 112.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5763-61-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5540 - 1.5580 @20?C
Appearance (Color)Clear, colourless to yellow
Formliquid
Assay (GC)> 96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only