trans-4-Aminocyclohexanol hydrochloride, 97%
trans-4-Aminocyclohexanol hydrochloride, 97%
trans-4-Aminocyclohexanol hydrochloride, 97%
trans-4-Aminocyclohexanol hydrochloride, 97%
Thermo Scientific Chemicals

trans-4-Aminocyclohexanol hydrochloride, 97%

CAS: 50910-54-8 | C6H14ClNO | 151.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25070.14
also known as B25070-14
Price (USD)
85.65
Online Exclusive
95.30
Save 9.65 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.30
Save 9.65 (10%)
Each
Add to cart
Chemical Identifiers
CAS50910-54-8
IUPAC Name4-aminocyclohexan-1-ol hydrochloride
Molecular FormulaC6H14ClNO
InChI KeyRKTQEVMZBCBOSB-UHFFFAOYSA-N
SMILESCl.NC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Water Content (Karl Fischer Titration)≤1.5%
Melting Point (clear melt)224.5-233.5?C
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only