2-Bromo-5-methylpyridine, 98+%
2-Bromo-5-methylpyridine, 98+%
2-Bromo-5-methylpyridine, 98+%
Thermo Scientific Chemicals

2-Bromo-5-methylpyridine, 98+%

CAS: 3510-66-5 | C6H6BrN | 172.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25083.22
also known as B25083-22
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
414.65
Online Exclusive
460.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS3510-66-5
IUPAC Name2-bromo-5-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyYWNJQQNBJQUKME-UHFFFAOYSA-N
SMILESCC1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown
FormCrystals or powder or crystalline powder or low melting crystalline solid
Assay (GC)≥98.0%
Melting Point (clear melt)40-47°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only