Trimethylsulfonium bromide, 98%
Trimethylsulfonium bromide, 98%
Trimethylsulfonium bromide, 98%
Trimethylsulfonium bromide, 98%
Thermo Scientific Chemicals

Trimethylsulfonium bromide, 98%

CAS: 3084-53-5 | C3H9BrS | 157.069 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B25098.22
also known as B25098-22
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 30-Jun-2026
161.00
Save 24.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3084-53-5
IUPAC Nametrimethylsulfanium bromide
Molecular FormulaC3H9BrS
InChI KeyGOTIICCWNAPLMN-UHFFFAOYSA-M
SMILES[Br-].C[S+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only