Potassium D-gluconate, 99%
Potassium D-gluconate, 99%
Potassium D-gluconate, 99%
Potassium D-gluconate, 99%
Thermo Scientific Chemicals

Potassium D-gluconate, 99%

CAS: 299-27-4 | C6H11KO7 | 234.245 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number B25135.0B
Price (USD)
180.65
Special offer
Online exclusive
Ends: 31-Dec-2025
212.00
Save 31.35 (15%)
Each
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)
180.65
Special offer
Online exclusive
Ends: 31-Dec-2025
212.00
Save 31.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS299-27-4
IUPAC Namepotassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular FormulaC6H11KO7
InChI KeyHLCFGWHYROZGBI-JJKGCWMISA-M
SMILES[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5% (dry wt. basis)
CommentReducing substances ≤ 1.0%
Loss on Drying≤ 3.0% (105?C, 4h)
View more
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium D-gluconate widely used as sequestering compounds for metal ions in alkaline systems. It is used in a newly developed process entitled ARS for the prevention of shrinkage in dyed wool fabric. Potassium gluconate has been shown to elevate lactate production, and ATP content, as well as induce the Hemoglobin (methemoglobin) reduction rate in rat erythrocytes. It is used in cell culture and in large scale applications such as the cleaning of metal surfaces.

Solubility
Soluble in water. (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

General References:

  1. Jeanette M. Cardamone; William C. Damert. Low-temperature Dyeing of Wool Processed for Shrinkage Control. Textile Research Journal.2006, 76, (1), 78-85.
  2. Rita Wegmüller; Fabian Tay; Christophe Zeder; Marica Brnic; Richard F Hurrell. Zinc absorption by young adults from supplemental zinc citrate is comparable with that from zinc gluconate and higher than from zinc oxide. Journal of Nutrition.2014, 144, (2), 132-136.