4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%
4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%
Thermo Scientific Chemicals

4-(4-Hydroxy-3-methoxyphenyl)-2-butanone, 97%

CAS: 122-48-5 | C11H14O3 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25150.06
also known as B25150-06
Price (USD)
52.65
Online Exclusive
58.60
Save 5.95 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
52.65
Online Exclusive
58.60
Save 5.95 (10%)
Each
Chemical Identifiers
CAS122-48-5
IUPAC Name4-(4-hydroxy-3-methoxyphenyl)butan-2-one
Molecular FormulaC11H14O3
InChI KeyOJYLAHXKWMRDGS-UHFFFAOYSA-N
SMILESCOC1=CC(CCC(C)=O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)35.5-44.5?C
Assay (GC)≥96.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only