2,3,5,6-Tetrafluorobenzyl alcohol, 98%
2,3,5,6-Tetrafluorobenzyl alcohol, 98%
2,3,5,6-Tetrafluorobenzyl alcohol, 98%
Thermo Scientific Chemicals

2,3,5,6-Tetrafluorobenzyl alcohol, 98%

CAS: 4084-38-2 | C7H4F4O | 180.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25162.14
also known as B25162-14
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
123.65
Online Exclusive
137.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS4084-38-2
IUPAC Name(2,3,5,6-tetrafluorophenyl)methanol
Molecular FormulaC7H4F4O
InChI KeyAGWVQASYTKCTCC-UHFFFAOYSA-N
SMILESOCC1=C(F)C(F)=CC(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to pale yellow
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Melting Point (clear melt)33.0-40.0?C
Refractive Index1.4560-1.4600 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only