4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Bromobenzaldehyde diethyl acetal, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
B25163.14
also known as B25163-14
25 g
Catalog number B25163.14
also known as B25163-14
Price (USD)
69.50
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
69.50
Each
Add to cart
Chemical Identifiers
CAS34421-94-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5115-1.5155 @ 20?C
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde diethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only
Donald R Gauthier; Ronald H Szumigala; Joseph D Armstrong III; R.P Volante. Stereoelectronic effects in the DIBAL reduction of aryl-1,2-ethanediol benzylidene acetals. Tetrahedron Letters.2001, 42, 7011-7014.