(R)-(-)-2-Amino-1-butanol, 98%
(R)-(-)-2-Amino-1-butanol, 98%
(R)-(-)-2-Amino-1-butanol, 98%
Thermo Scientific Chemicals

(R)-(-)-2-Amino-1-butanol, 98%

CAS: 5856-63-3 | C4H11NO | 89.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25212.22
also known as B25212-22
Price (USD)
506.65
Special offer
Online exclusive
Ends: 30-Jun-2026
596.00
Save 89.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
506.65
Special offer
Online exclusive
Ends: 30-Jun-2026
596.00
Save 89.35 (15%)
Each
Chemical Identifiers
CAS5856-63-3
IUPAC Name2-aminobutan-1-ol
Molecular FormulaC4H11NO
InChI KeyJCBPETKZIGVZRE-UHFFFAOYNA-N
SMILESCCC(N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Optical Rotation-6.5° to -10.5° (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only