2-Ethyl-6-methylphenol, 97%
2-Ethyl-6-methylphenol, 97%
2-Ethyl-6-methylphenol, 97%
Thermo Scientific Chemicals

2-Ethyl-6-methylphenol, 97%

CAS: 1687-64-5 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25213.03
also known as B25213-03
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.70
Save 6.05 (15%)
Each
Chemical Identifiers
CAS1687-64-5
IUPAC Name2-ethyl-6-methylphenol
Molecular FormulaC9H12O
InChI KeyCIRRFAQIWQFQSS-UHFFFAOYSA-N
SMILESCCC1=CC=CC(C)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Refractive Index1.5275-1.5325 @ 20?C
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only