2-Ethyl-6-methylphenol, 97%
2-Ethyl-6-methylphenol, 97%
2-Ethyl-6-methylphenol, 97%
Thermo Scientific Chemicals

2-Ethyl-6-methylphenol, 97%

CAS: 1687-64-5 | C9H12O | 136.194 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25213.14
also known as B25213-14
Price (USD)
438.65
Online Exclusive
487.00
Save 48.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
438.65
Online Exclusive
487.00
Save 48.35 (10%)
Each
Chemical Identifiers
CAS1687-64-5
IUPAC Name2-ethyl-6-methylphenol
Molecular FormulaC9H12O
InChI KeyCIRRFAQIWQFQSS-UHFFFAOYSA-N
SMILESCCC1=CC=CC(C)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5275-1.5325 @ 20?C
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only