Itaconic anhydride, 97%
Itaconic anhydride, 97%
Itaconic anhydride, 97%
Thermo Scientific Chemicals

Itaconic anhydride, 97%

CAS: 2170-03-8 | C5H4O3 | 112.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B25216.14
also known as B25216-14
Price (USD)
36.65
Online Exclusive
40.30
Save 3.65 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.30
Save 3.65 (9%)
Each
Chemical Identifiers
CAS2170-03-8
IUPAC Name3-methylideneoxolane-2,5-dione
Molecular FormulaC5H4O3
InChI KeyOFNISBHGPNMTMS-UHFFFAOYSA-N
SMILESC=C1CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)65.0-72.0?C
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Itaconic anhydride is used in immobilization of β-glucosidase. It undergoes estrification to obtain suitable monomers for emulsion polymerization itaconates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
RUO – Research Use Only

General References:

  1. Tapas Palai; Ashok Kumar; Prashant K. Bhattacharya. Kinetic studies and model development for the formation of galacto-oligosaccharides from lactose using synthesized thermo-responsive bioconjugate. Enzyme and Microbial Technology.2009, 70,
  2. L. Fischer; F. Peibker. A covalent two-step immobilization technique using itaconic anhydride. Applied Microbiology and Biotechnology.1998, 49, 129-135.