2-Bromo-4'-methylacetophenone, 98%
2-Bromo-4'-methylacetophenone, 98%
2-Bromo-4'-methylacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-4'-methylacetophenone, 98%

CAS: 619-41-0 | C9H9BrO | 213.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25217.14
also known as B25217-14
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
153.65
Online Exclusive
171.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS619-41-0
IUPAC Name2-bromo-1-(4-methylphenyl)ethan-1-one
Molecular FormulaC9H9BrO
InChI KeyKRVGXFREOJHJAX-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale green
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)49.0-56.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only