6,7-Dimethoxy-1-tetralone, 97%
6,7-Dimethoxy-1-tetralone, 97%
6,7-Dimethoxy-1-tetralone, 97%
Thermo Scientific Chemicals

6,7-Dimethoxy-1-tetralone, 97%

CAS: 13575-75-2 | C12H14O3 | 206.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25226.14
also known as B25226-14
Price (USD)
1,034.65
Online Exclusive
1,149.00
Save 114.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
1,034.65
Online Exclusive
1,149.00
Save 114.35 (10%)
Each
Chemical Identifiers
CAS13575-75-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
Melting Point (clear melt)94.5-103.5?C
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only