1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-phenylbutane is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Manabu Horikawa; Kazuhiro Tateda; Etsu Tuzuki; Yoshikazu Ishii; Chihiro Ueda; Tohru Takabatake; Shinichi Miyairi; Keizou Yamaguchi; Masaji Ishiguro. Synthesis of Pseudomonas quorum-sensing autoinducer analogs and structural entities required for induction of apoptosis in macrophages. Bioorganic & Medicinal Chemistry Letters. 2006, 16,(8), 2130-2133.
- Richard A.Glennon; John J.Salley Jr.; Odd S.Steinsland; Sharon Nelson. Synthesis and evaluation of novel alkylpiperazines as potential dopamine antagonists. J. Med. Chem. 1981, 24,(6), 678-683.