2-Bromo-4-methylpyridine, 96%
2-Bromo-4-methylpyridine, 96%
2-Bromo-4-methylpyridine, 96%
Thermo Scientific Chemicals

2-Bromo-4-methylpyridine, 96%

CAS: 4926-28-7 | C6H6BrN | 172.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25253.06
also known as B25253-06
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.90
Save 10.25 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 30-Jun-2026
70.90
Save 10.25 (14%)
Each
Chemical Identifiers
CAS4926-28-7
IUPAC Name2-bromo-4-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyLSZMVESSGLHDJE-UHFFFAOYSA-N
SMILESCC1=CC=NC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5605-1.5665 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only