3-Nitrophthalonitrile, 99%
3-Nitrophthalonitrile, 99%
3-Nitrophthalonitrile, 99%
3-Nitrophthalonitrile, 99%
Thermo Scientific Chemicals

3-Nitrophthalonitrile, 99%

CAS: 51762-67-5 | C8H3N3O2 | 173.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25269.14
also known as B25269-14
Price (USD)
192.65
Special offer
Online exclusive
Ends: 30-Jun-2026
227.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
192.65
Special offer
Online exclusive
Ends: 30-Jun-2026
227.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS51762-67-5
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Melting Point (clear melt)158.0-165.0°C
FormPowder
Identification (FTIR)Conforms
Appearance (Color)White to pale cream to pale yellow to black
3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophthalonitrile is a useful dye for biological research purposes. It is used as an intermediate in the manufacture of pharmaceuticals, chemicals, dyestuff and phthalocyanine blue pigments.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incombatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Sivanesan, A.; Adamkiewicz, W.; Kalaivani, G.; Kaminska, A.; Waluk, J.; Holyst, R.; Izake, E. L. Towards improved precision in the quantification of surface-enhanced Raman scattering (SERS) enhancement factors: a renewed approach. Analyst 2015, 140 (2), 489-496.
  2. Zhou, Y.; Guofei, G.; Zhao, H.; Song, L.; Fang, X. Synthesis and properties of transparent polyimides derived from trans-1,4-bis(2,3-dicarboxyphenoxy)cyclohexane dianhydride. RSC Adv. 2015, 5 (66), 53926-53934.