Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%
Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%
Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%
Thermo Scientific Chemicals

Copper(II) hexafluoro-2,4-pentanedionate hydrate, 98%

CAS: 155640-85-0 | C10H2CuF12O4 | 477.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25284.03
also known as B25284-03
Price (USD)
25.60
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
25.60
Each
Chemical Identifiers
CAS155640-85-0
IUPAC Namecopper(2+) bis((2Z)-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Molecular FormulaC10H2CuF12O4
InChI KeyHZXGNBMOOYOYIS-PAMPIZDHSA-L
SMILES[Cu++].[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F.[O-]\C(=C/C(=O)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Iodometric Titration)≥97.5 to ≤102.5% (dry weight basis)
Melting Point100-134?C
Water Content (Karl Fischer Titration)≤8.0%
FormPowder
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only