Ethyl phenylglyoxylate, 98%
Ethyl phenylglyoxylate, 98%
Ethyl phenylglyoxylate, 98%
Thermo Scientific Chemicals

Ethyl phenylglyoxylate, 98%

CAS: 1603-79-8 | C10H10O3 | 178.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25292.06
also known as B25292-06
Price (USD)
30.65
Online Exclusive
33.50
Save 2.85 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.50
Save 2.85 (9%)
Each
Chemical Identifiers
CAS1603-79-8
IUPAC Nameethyl 2-oxo-2-phenylacetate
Molecular FormulaC10H10O3
InChI KeyQKLCQKPAECHXCQ-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5135-1.5175 @ 20°C
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.

Solubility
Soluble in water 1143 mg/L @ 25°C .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Kiyoshi Tanaka; Tomoya Okada; Fumio Yoneda; Tomohisa Nagamatsu; Kazunori Kuroda. Preparation of chiral 5-deazaflavin derivatives and their asymmetric reduction of ethyl benzoylformate. Tetrahedron Letters. 1984, 25 (16), 1741-1742.
  2. Mária Sutyinszki; Kornél Szöri; Károly Felföldi; Mihály Bartók. 98% Enantioselectivity in the asymmetric synthesis of a useful chiral building block by heterogeneous method: Enantioselective hydrogenation of ethyl-benzoylformate over cinchona modified Pt/Al2O3 catalysts in the acetic acid. Catalysis Communications. 2002, 3 (3), 125-127.