Methyl 4-iodosalicylate, 98%
Methyl 4-iodosalicylate, 98%
Methyl 4-iodosalicylate, 98%
Methyl 4-iodosalicylate, 98%
Thermo Scientific Chemicals

Methyl 4-iodosalicylate, 98%

CAS: 18179-39-0 | C8H7IO3 | 278.045 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25438.14
also known as B25438-14
Price (USD)
163.00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
163.00
Each
Add to cart
Chemical Identifiers
CAS18179-39-0
IUPAC Namemethyl 2-hydroxy-4-iodobenzoate
Molecular FormulaC8H7IO3
InChI KeyWUFUURSWOJROKY-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(O)C=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)67.5-73.5°C
Appearance (Color)White to dark yellow to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only