3-Ethoxymethylene-2,4-pentanedione, 97+%
3-Ethoxymethylene-2,4-pentanedione, 97+%
3-Ethoxymethylene-2,4-pentanedione, 97+%
Thermo Scientific Chemicals

3-Ethoxymethylene-2,4-pentanedione, 97+%

CAS: 33884-41-2 | C8H12O3 | 156.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25445.14
also known as B25445-14
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS33884-41-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Refractive Index1.4835-1.4875 @ 20?C
Appearance (Color)Clear pale yellow to yellow to orange
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only