(+/-)-Indoline-2-carboxylic acid, 95%
(+/-)-Indoline-2-carboxylic acid, 95%
(+/-)-Indoline-2-carboxylic acid, 95%
Thermo Scientific Chemicals

(+/-)-Indoline-2-carboxylic acid, 95%

CAS: 78348-24-0 | C9H8NO2 | 162.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25450.06
also known as B25450-06
Price (USD)
58.65
Online Exclusive
65.40
Save 6.75 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
65.40
Save 6.75 (10%)
Each
Chemical Identifiers
CAS78348-24-0
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.5%
Optical Rotation0.0 ± 0.2? (C=1 in 1N HCl)
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Assay (Silylated GC)≥94.0%
Appearance (Color)Pale cream to pale brown
View more
(±)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Yong Hae Kim; Sung Han Kim; Doo Han Park. Asymmetric 1,3-dipolar cycloaddition of nitrile oxides to new chiral acrylamides derived from (S)-indoline-2-carboxylic acid. Tetrahedron Letters. 1993, 34,(38), 6063-6066.
  2. Jin-Qiang Liu; Xin-Xiang Cao; Baoming Ji; Bangtun Zhao. Determination and correlation of solubilities of (S)-indoline-2-carboxylic acid in six different solvents from (283.15 to 358.15) K. J. Chem. Eng. Data. 2013, 58,(9), 2414-2419.